Disposable Dissection Boards
About this item
The DispoCut™ disposable dissection board is strong but inexpensive enough to throw away. It is a clean, safer and more efficient way of handling infectious tissue specimens.
- White, cut-resistant boards with dimensional scalles
- Reversible, printed on both sides, Disposable
- Extruded co-polymer resin boards
- Printed imperial and metric dimensional scales in inches, centimeters and millimeters
- Seven dimensional circles from 4 to 16 mm and five larger ones from 2 to 6 cm in Ø
- Strong surface will not dull knifes but well suited for using dissecting pins
- Temperature performance range: −27 to +71 °C (−17 to +160 °F)
- Melting point: 162 °C (324 °F)
Ideal for pathology labs, medical schools, classroom dissections, forensic labs, medical research labs, pharmaceutical labs, surgery and more. When sterilizing, DispoCut™ may be safely wiped down with alcohol or autoclaved.
Applications: Dissection boards for use pathology labs, schools, forensic labs and more.
3 Options Available Below
Product Details & Documents
The DispoCut™ disposable dissection board is strong but inexpensive enough to throw away. It is a clean, safer and more efficient way of handling infectious tissue specimens.
- White, cut-resistant boards with dimensional scalles
- Reversible, printed on both sides, Disposable
- Extruded co-polymer resin boards
- Printed imperial and metric dimensional scales in inches, centimeters and millimeters
- Seven dimensional circles from 4 to 16 mm and five larger ones from 2 to 6 cm in Ø
- Strong surface will not dull knifes but well suited for using dissecting pins
- Temperature performance range: −27 to +71 °C (−17 to +160 °F)
- Melting point: 162 °C (324 °F)
Ideal for pathology labs, medical schools, classroom dissections, forensic labs, medical research labs, pharmaceutical labs, surgery and more. When sterilizing, DispoCut™ may be safely wiped down with alcohol or autoclaved.
Applications: Dissection boards for use pathology labs, schools, forensic labs and more.
Product Family Options
Product Information
- ColorMaterialL×WAvailabilityPrice
- 77666-350DispoCut dissection board, small, case of 96DispoCut dissection board, small, case of 96WhiteCopolymer resin6×8"Log in for availabilityCase of 96$810.54Supplier Number: WP-630-1
- 77666-352DispoCut dissection board, medium, case of 48DispoCut dissection board, medium, case of 48WhiteCopolymer resin9×12"Log in for availabilityCase of 48$600.40Supplier Number: WP-630-2
- 77666-354DispoCut dissection board, large, case of 24DispoCut dissection board, large, case of 24WhiteCopolymer resin12×19"Log in for availabilityCase of 24$595.66Supplier Number: WP-630-3
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



