About this item
For digestion of native collagen fibrils.
- Compatible with a variety of cell tissue types
Digest native collagen fibrils in connective tissues with Collagenase Type I. Sourced from Clostridium histolyticum, this enzymatic preparation contains the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin. Collagenase Type I has been used for the digestion of many human tissues and cell types, including the intestine (Barthel et al.), mammary glands (Huss and Kratz), prostate (Le et al.), endothelial cells (Ganguly et al.), and dorsal root ganglia (Dib-Hajj et al.).
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Species:HumanMouseNon-human primateRat