NeuroCult™ Enzymatic Dissociation Kit for Adult CNS Tissue (Mouse and Rat)
Catalogue # MSPP-5715
Fournisseur: Stemcell Technologies
Numéro CAS:
Spécifications
- Species:MouseRat
- Description:NeuroCult™ enzymatic dissociation kit for adult CNS tissue
- Size:1 kit
- Cat. no.:MSPP-5715
Spécifications
À propos de cet article
Kit for enzymatic dissociation of adult mouse and rat CNS tissue.
- Obtain cleaner cultures with a ready-to-use solution
- Achieve fast and reproducible enzymatic dissociation
- Produce higher cell number yields and increased viability
Conduct reliable and efficient digestion and dissociation procedures for mouse and rat central nervous system (CNS) tissue with ready-to-use NeuroCult™ Enzymatic Dissociation Kit. Optimized to ensure a rapid and reproducible procedure, this kit for produces a single-cell suspension that is ready for immediate use in downstream applications and yields high cell numbers and viabilities.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').