Order Entry
Canada
ContactUsLinkComponent
ACCUMAX™ Cell Detachment Solution
ACCUMAX™ Cell Detachment Solution
Catalog # MSPP-07921
CAS Number:  
undefined
ACCUMAX™ Cell Detachment Solution
Catalog # MSPP-07921
Supplier Number:  07921
CAS Number:  

Specifications

  • Species:
    Human
    Mouse
    Non-human primate
    Rat
  • Description:
    ACCUMAX™ cell detachment solution
  • Size:
    100 ml
  • Cat. no.:
    MSPP-07921

Specifications

About this item

Cell detachment solution.

  • Increase the accuracy of cell counts, using manual or automated methods
  • Effectively dissolve cell clumps, such as spheroids and neurospheres
  • Obtain cleaner cultures with a ready-to-use solution, free from mammalian and bacterial-derived products

Generate single-cell suspensions from clumped cell cultures for accurate cell counting with ACCUMAX™. This ready-to-use solution of proteolytic and collagenolytic enzymes effectively dissolves cell clumps and effectively facilitates the detachment of cells from primary tissue, as well as standard and adhesion-coated culture plasticware.

ACCUMAX™ is free of mammalian- or bacterial- derived products to ensure reproducible and reliable experiments, and has been shown to work effectively across a wide variety of cell types. Each lot of ACCUMAX™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromogenic tetrapeptides), and cell detachment from tissue culture plastic.

Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').