Order Entry
ContactUsLinkComponent
94 results for Shakers

You searched for: Rotating/Rocking Shakers

Rotating/Rocking Shakers

Rotating and rocking shakers combine the functions of a laboratory rocker with a shaker. Complete with variable speed settings and a configurable tilt angle, these compact instruments tailor operations for blot washing, gel staining, and hybridization applications. The rotating and rocking units can be placed inside incubators or cold rooms as required. Rotating and rocking shaker options offer single- or double-tiered platforms that are durably constructed and easy-to-clean.

Sort by
VWR® Tube Rotators

VWR® Tube Rotators

Supplier: VWR International

VWR® Tube rotators are ideal for applications requiring highly efficient and consistent mixing. VWR offers rugged and reliable fixed speed, digital and large capacity models.

Expand 3 Items
Loading...
VWR® Signature™ 3D Rotator Waver, 230 V

VWR® Signature™ 3D Rotator Waver, 230 V

Supplier: VWR International

3-D rotator waver is ideal for blood samples, DNA extractions, blotting techniques, and general mixing of various size tubes.

Expand 1 Items
Loading...
VWR® Variable Speed Rocker

VWR® Variable Speed Rocker

Supplier: VWR International

Designed to provide precise speed and tilt angles for a broad range of molecular and biological mixing applications.

Expand 1 Items
Loading...
VWR® Tube Rocker Collection

VWR® Tube Rocker Collection

Supplier: VWR International

The VWR® Tube Rocker Collection features Tube Rockers available in a "Mini" size as well as an "XL" size and in Fixed Speed or a Variable Speed models.

Expand 3 Items
Loading...
VWR® Nutating Mixer

VWR® Nutating Mixer

Supplier: VWR International

The three-dimensional shaking motion of this mixer is gentle yet efficient, making it ideal for mixing samples in blood and centrifuge tubes.

Expand 1 Items
Loading...
VWR® Analog Rocking Platform Shakers

VWR® Analog Rocking Platform Shakers

Supplier: VWR International

VWR® Analog Rocking Platform Shakers

Expand 2 Items
Loading...
VWR® Signature™ Rocking Platform Shaker, 230 V

VWR® Signature™ Rocking Platform Shaker, 230 V

Supplier: VWR International

Ideal for cell culture work, staining and destaining gels, hybridization procedures, hematology, and blotting techniques.

Expand 1 Items
Loading...
VWR® Mini Tube Rotator

VWR® Mini Tube Rotator

Supplier: VWR International

The VWR® Mini Tube Rotator provides a wide range of mixing capabilities suitable for an array of laboratory applications.

Expand 7 Items
Loading...
VWR® Mini Tube Rotator, 230 V

VWR® Mini Tube Rotator, 230 V

Supplier: VWR International

The VWR® Mini Tube Rotator provides a wide range of mixing capabilities suitable for an array of laboratory applications.

Expand 1 Items
Loading...
VWR® 2D Platform Rockers

VWR® 2D Platform Rockers

Supplier: VWR International

VWR® 2D platform rockers are ideal for use in blotting and staining applications as well as gentle mixing of solutions and reagents.

Expand 2 Items
Loading...
VWR® 3D Nutating Mixers

VWR® 3D Nutating Mixers

Supplier: VWR International

VWR® 3D nutating mixers are designed for sample mixing with combined motions of an orbital shaker and 2D platform rocker. These mixers are well suited for many routine laboratory applications that require thorough sample mixing.

Expand 4 Items
Loading...
VWR® Signature™ 3D Rotator Waver

VWR® Signature™ 3D Rotator Waver

Supplier: VWR International

3-D rotator waver is ideal for blood samples, DNA extractions, blotting techniques, and general mixing of various size tubes.

Expand 2 Items
Loading...
VWR® Signature™ Rocking Platform Shaker

VWR® Signature™ Rocking Platform Shaker

Supplier: VWR International

Ideal for cell culture work, staining and destaining gels, hybridization procedures, hematology, and blotting techniques.

Expand 2 Items
Loading...
VWR® Analog 3D Waving Platform Shaker, 230 V

VWR® Analog 3D Waving Platform Shaker, 230 V

Supplier: VWR International

VWR® Analog waving platform shakers are an easy and economical option for all of your waving shaking needs.

Expand 1 Items
Loading...
VWR® Rotating Mixer XL

VWR® Rotating Mixer XL

Supplier: VWR International

The VWR® rotating mixer XL mixes common laboratory samples gently or vigorously.

Expand 1 Items
Loading...
VWR® Tube Roller Series

VWR® Tube Roller Series

Supplier: VWR International

VWR® tube rollers are designed for gentle yet complete mixing of a wide range of sample vessels and tubes.

Expand 2 Items
Loading...
VWR® Tube Revolvers

VWR® Tube Revolvers

Supplier: VWR International

Efficient, powerful tube revolvers fixed or variable speed, with 5 interchangeable rotor series.

Expand 2 Items
Loading...
VWR® Tube Rotator and Rotisseries

VWR® Tube Rotator and Rotisseries

Supplier: VWR International

The unique design of the VWR® Tube Rotator allows for a variety of tube sizes and mixing applications to be performed efficiently on one instrument.

Expand 1 Items
Loading...
VWR® Analog 3D Waving Platform Shaker

VWR® Analog 3D Waving Platform Shaker

Supplier: VWR International

VWR® Analog waving platform shakers are an easy and economical option for all of your waving shaking needs.

Expand 1 Items
Loading...
Rotators for Cell Culture, Ace Glass

Rotators for Cell Culture, Ace Glass

Supplier: Ace Glass

This variable speed rotator is driven by a continuous-duty motor that can withstand a demanding schedule and long hours of operation.

Expand 1 Items
Loading...
Corning® LSE™ Nutating Mixer, Corning

Corning® LSE™ Nutating Mixer, Corning

Supplier: Corning

The LSE Nutating mixer is a small personal sized 3D mixer which is ideal for ambient and cold room applications for example immunohistochemistry, mixing and resupending small containers such as blood and centrifuge tubes.

Expand 1 Items
Loading...
Rotators for Cell Culture, 240 V

Rotators for Cell Culture, 240 V

Supplier: Ace Glass

This variable speed rotator is driven by a continuous-duty motor that can withstand a demanding schedule and long hours of operation.

Expand 2 Items
Loading...
Corning® LSE™ Platform Rockers, 230 V

Corning® LSE™ Platform Rockers, 230 V

Supplier: Corning

The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications.

Expand 2 Items
Loading...
Corning® LSE™ Platform Rockers, Corning

Corning® LSE™ Platform Rockers, Corning

Supplier: Corning

The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications.

Expand 2 Items
Loading...
CELLSPIN Spinner System, Argos Technologies

CELLSPIN Spinner System, Argos Technologies

Supplier: Argos Technologies

CELLSPIN a reliable cell culture system.

Expand 12 Items
Loading...

Digital Mini Rotators, Thermo Scientific

Supplier: Thermo Fisher Scientific

The Thermo Scientific™ Digital mini rotator has been designed for culturing bacteria and fungi, hybridization assays, and gel staining and de-staining.

Expand 1 Items
Loading...
Digital Platform Rocker, Thermo Scientific

Digital Platform Rocker, Thermo Scientific

Supplier: Thermo Fisher Scientific

The Digital Platform Rocker's double platform accommodates more samples while providing consistent rocking motion and easy angle adjustments.

Expand 1 Items
Loading...
Digital Tube Rocker, Thermo Scientific

Digital Tube Rocker, Thermo Scientific

Supplier: Thermo Fisher Scientific

Digital Tube Rocker is a precision-controlled platform mixer that accommodates double capacity in a compact footprint for smooth rocking and uniform suspension.

Expand 1 Items
Loading...
Tube Revolver/Rotator, Thermo Scientific

Tube Revolver/Rotator, Thermo Scientific

Supplier: Thermo Fisher Scientific

Tube Revolver; Global version; Variable Speed; Digital Display; 10-40rpm; 100-240V, 50/60Hz; adaptor for multi plugs,7.7x6x8.9in,Capacity Combination of four paddles of: 7x 10/15mL tubes, 7 x 5/7mL tubes, 15 x 1.5mL/2mL tubes, 21 x 0.5/0.8mL tubes, One paddle of 6 x 50mL tubes

Expand 2 Items
Loading...

Digital Mini Rotators, 230 V

Supplier: Thermo Fisher Scientific

The Thermo Scientific™ Digital mini rotator has been designed for culturing bacteria and fungi, hybridization assays, and gel staining and de-staining.

Expand 1 Items
Loading...
Sort by
Recommended for You