Orders LinkContactUsLinkComponent
McPherson-Vannas Scissors
McPherson-Vannas Scissors
McPherson-Vannas Scissors
Certificates
McPherson-Vannas Scissors
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made up of stainless steel.
  • 0.3 mm rounded tips
Product Family Options
Product Information
  • Length
    Blade length
    Availability
    Price
  • 77663-140
    McPherson-Vannas scissors, 0.22 mm tips, straight
    McPherson-Vannas scissors, 0.22 mm tips, straight
    8 cm (3")
    3 mm
    Log in for availability
    Log in for availability
    Each
    Each - $622.85
    $622.85
    Each
       $622.85
    Supplier Number: 503215
  • 77663-138
    McPherson-Vannas scissors, 0.22 mm tips, 45° angled to side
    McPherson-Vannas scissors, 0.22 mm tips, 45° angled to side
    8 cm (3")
    5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $579.05
    $579.05
    Each
       $579.05
    Supplier Number: 503214
  • 77663-144
    McPherson-Vannas scissors, 0.30 mm tips, curved
    McPherson-Vannas scissors, 0.30 mm tips, curved
    8 cm (3")
    5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $622.85
    $622.85
    Each
       $622.85
    Supplier Number: 503217
  • 77663-142
    McPherson-Vannas scissors, 0.50 mm tips, angled on flat
    McPherson-Vannas scissors, 0.50 mm tips, angled on flat
    8 cm (3")
    5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $622.85
    $622.85
    Each
       $622.85
    Supplier Number: 503216
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)