Student Micro Surgery Scissors
Product Details & Documents
Our economy line of spring scissors are perfect for students who are doing small animal surgeries or dissection in a research laboratory. Student spring scissors are manufactured from hardened surgical grade stainless steel. They are perfect for classroom use or as disposable instruments.
- 150 mm long
- 4 mm cuting edge
Specifications
- Blade type:Straight blades
- Handle:Flat handle
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)