$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Buffers
Biological Buffers
100 mM Sodium Phosphate with 10 mM EDTA and 0.01% Tween-20
100 mM Sodium Phosphate with 10 mM EDTA and 0.01% Tween-20
CA77442-678
:
Teknova
New Product
:
100 mM Sodium Phosphate with 10 mM EDTA and 0.01% Tween-20
CA77442-678
:
Teknova
:
S2216
New Product
:
Description:
100 mM sodium phosphate with 10 mM EDTA and 0.01% Tween-20
Size:
1000 ml
Sterility:
Sterile
Cat. no.:
CA77442-678
pH:
7.4