Orders LinkContactUsLinkComponent
Round Hollow Handled Forceps
Round Hollow Handled Forceps
Round Hollow Handled Forceps
Certificates
Round Hollow Handled Forceps
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Hollowing out the handles of long, round-handled instruments decreases the overall weight. This makes them efficient and non-fatiguing when operating under a microscope. Made from high quality stainless steel, these instruments are an excellent addition to a microsurgical instrument kit.
  • 15 cm (6") long
  • Round, hollow handles
Product Family Options
Product Information
  • Tip length
    Tip style
    Availability
    Price
  • 0.15 mm
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $267.65
    $267.65
    Each
       $267.65
    Supplier Number: 14113
  • 0.15 mm
    Straight, German
    Log in for availability
    Log in for availability
    Each
    Each - $391.82
    $391.82
    Each
       $391.82
    Supplier Number: 14113-G
  • 0.3 mm
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $262.79
    $262.79
    Each
       $262.79
    Supplier Number: 14114
  • 0.15 mm
    45° Angle
    Log in for availability
    Log in for availability
    Each
    Each - $352.81
    $352.81
    Each
       $352.81
    Supplier Number: 14209
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.