Order Entry
Canada
ContactUsLinkComponent
1M Sodium Phosphate Buffer
1M Sodium Phosphate Buffer
Catalog # CA77442-714
Supplier:  Teknova
New Product
CAS Number:  
1M Sodium Phosphate Buffer
Catalog # CA77442-714
Supplier:  Teknova
Supplier Number:  P3072
New Product
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Description:
    1M sodium phosphate buffer
  • Size:
    1000 ml
  • Sterility:
    Sterile
  • Cat. no.:
    CA77442-714
  • pH:
    7.2

Specifications

About this item

312 mM sodium phosphate monobasic, 688 mM sodium phosphate dibasic.