Vacu-vials® Copper Test Kit for use with Spectrophotometer
Catalog # 77674-766
Supplier: CHEMetrics
New Product
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Cat. no.:77674-766
- Range:0 - 12.00 ppm
- No. of Tests:30
Specifications
About this item
The method measures total soluble copper as ppm (mg/L) Cu. The test kits are applicable for analysis of drinking water, surface waters, groundwater, wastewater and seawater.
- Premixed
- Premeasured
- Precise
Ordering information: This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: Thirty ampoules, 25 ml sample cup, ampoule blank, and instructions.