Orders LinkContactUsLinkComponent
Microelectrode Holders
Microelectrode Holders
Microelectrode Holders
Certificates
Microelectrode Holders
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
WPI™s microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers.
  • Female connector
  • Pellet half-cell
  • No pressure port
  • Screw cap
  • With 6.3 mm Ø
A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps).
Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. The holder style selected will depend on the experimental application, space, and instrumentation.
 : All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
Product Family Options
Product Information
  • Availability
    Price
  • Log in for availability
    Log in for availability
    Each
    Each - $271.76
    $271.76
    Each
       $271.76
    Supplier Number: MEH3F4510
    Specifications:
    More
    Specifications
    Cat. No.
    77664-674
  • Log in for availability
    Log in for availability
    Each
    Each - $271.76
    $271.76
    Each
       $271.76
    Supplier Number: MEH3F4515
    Specifications:
    More
    Specifications
    Cat. No.
    77664-676
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)