Planet-Safe® Sampling Spoons
About this item
Made of PLA, a 100% planet-based bio-polymer, sustainabley derived from renewable resources.
- Proprietary formulation won’t warp in boiling liquids
- 10 ml level capacity
- Individually wrapped and sterile
Sit-flat foot keeps spoons level during weighing. Rigid construction allows easy break up of caked powders. Safe to incinerate and commercially compostable.
2 Options Available Below
Product Details & Documents
Made of PLA, a 100% planet-based bio-polymer, sustainabley derived from renewable resources.
- Proprietary formulation won’t warp in boiling liquids
- 10 ml level capacity
- Individually wrapped and sterile
Sit-flat foot keeps spoons level during weighing. Rigid construction allows easy break up of caked powders. Safe to incinerate and commercially compostable.
Product Family Options
Product Information
- LengthColorCapacityAvailabilityPrice
- Supplier Number: BPSP-1000Specifications:
More
Specifications
Cat. No.77674-450 - Supplier Number: BPSP-2000Specifications:
More
Specifications
Cat. No.77674-452