ContactUsLinkComponent
Stainless Steel Slanted Storage Rack, Bandy
  24450-010
 :  Bandy
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Stainless Steel Slanted Storage Rack, Bandy
  24450-010
 :  Bandy
 :  WRK60003
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
  • Load Capacity:
    800 lbs.
  • Cat. no.:
    24450-010
  • Supplier No.:
    WRK60003
Storage rack features rod-top shelves sloped at an 8° angle for safe storage of work-in-process
Constructed of 16-gauge, type 304 stainless steel, completely electropolished. Adjustable leveling feet.