MesenCult™ SF Attachment Substrate
Catalog # MSPP-05424
Supplier: Stemcell Technologies
CAS Number:
Specifications
- Description:Cult™-SF Attachment substrate
- Size:5 mg
- Cat. no.:MSPP-05424
Specifications
About this item
Serum-free attachment substrate for human mesenchymal stem cells - 5 mg.
Support the attachment and growth of human mesenchymal stem and progenitor cell (MSC) cultures in vitro with MesenCult™-SF Attachment Substrate. This serum-free cell culture substrate is optimized to enhance the expansion of human MSCs in vitro and support their enumeration using the colony-forming unit–fibroblast (CFU-F) assay.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further re sale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet ( collectively, the 'Product Restrictions').