LED Line Projectors, 80 W
New Product
Mark the unmarkable with LED Line projectors from Brady. Ensure safety and showcase professionalism anywhere with projectors ideal for challenging and changing environments.
- Projects up to 48'
- Narrow, medium or wide projector lens
- 80 W LED lamp
- Two-year warranty
- 110 to 240 V power supply voltage
- 3 to 6" projection width range
Identify surfaces where paint and tape can not, including those that are hot, humid, dusty, grimy, uneven and irregular. Eliminate the prep, install, waste and training needed with disposable tapes and paints. Maintenance-free operation thanks to no fans, motors or moving parts. Waterproof design is safe for washdowns. Designed for general industrial environments. Wide temperature and power range. Line length reaches up to 36 feet and line width spans 4 to 6 inches. Select lens based on line width needs: 53° (narrow), 73° (medium) or 90° (wide).
: Also includes mounting yoke and power cord.
Frequently Bought Together