Order Entry
Canada
ContactUsLinkComponent
illustra NICK Columns, Cytiva
illustra NICK Columns, Cytiva
Supplier:  Cytiva
Certificates
undefined
illustra NICK Columns, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Scalable
  • Refine target molecule:
    DNA preps
  • Sample type:
    DNA preps
  • Target molecule:
    DNA
  • Storage conditions:
    4 to 30°C
  • Sample size:
    100 µL
  • Binding capacity:
    100 µg

Specifications

Frequently Bought Together