Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ release human PE positive selection kit
- Cat. no.:MSPP-17654
Specifications
About this item
Immunomagnetic positive selection of particle-free human cells labeled with PE-conjugated antibodies.
- Highly purified cells labeled with PE-conjugated antibodies isolated from human or mouse tissues in less than 40 minutes
- No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Human PE Positive Selection Kit is designed to isolate cells that are labeled with PE-conjugated antibodies from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection.
Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated, PE-antibody labeled cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands. This product replaces the EasySep™ Human PE Positive Selection Kit, providing highly purified particle-free cells.