ContactUsLinkComponent
EasySep™ HLA Chimerism Whole Blood CD4 Positive Selection Kits
EasySep™ HLA Chimerism Whole Blood CD4 Positive Selection Kits
Catalog # MSPP-17888
undefined
EasySep™ HLA Chimerism Whole Blood CD4 Positive Selection Kits
Catalog # MSPP-17888
Supplier Number:  17888
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Capacity:
    60 ml whole blood processed
  • Description:
    EasySep™ HLA chimerism whole blood CD4 positive selection kit
  • Cat. no.:
    MSPP-17888
Specifications
About this item
Column-free immunomagnetic positive selection of human CD4+ cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ HLA Chimerism Whole Blood CD4 Positive Selection is designed to isolate CD4+ cells from fresh whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD4 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD4 antigen is expressed on helper T cells and more weakly on monocytes and tissue macrophages.
This product replaces the EasySep™ Human Whole Blood CD4 Positive Selection Kit for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').