ContactUsLinkComponent
EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kits
EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kits
Catalog # MSPP-18782
undefined
EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kits
Catalog # MSPP-18782
Supplier Number:  18782
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ mouse CD25 regulatory T cell positive selection kit
  • Cat. no.:
    MSPP-18782
Specifications
About this item
Immunomagnetic positive selection of mouse regulatory T cells.
  • Fast and easy-to-use
  • Up to 93% purity
  • No columns required
The EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kit is designed to isolate highly purified CD25+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. The kit targets CD25 regulatory T cells for positive selection with a PE-labeled antibody directed against the CD25 surface marker. Desired cells are labeled with antibodies and magnetic particles. The cells are separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').