If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ release human PSC-derived neural crest cell positive selection kit
- Cat. no.:MSPP-100-0047
Specifications
About this item
Immunomagnetic positive selection kit using particle release technology
- Highly purified human PSC-derived neural crest cells isolated in less than 20 minutes
- No-wash removal of EasySep™ Releasable RapidSpheres™
- Compatible with downstream differentiation protocols
The EasySep™ Release Human PSC-Derived Neural Crest Cell Positive Selection Kit is designed to isolate CD271⁺ cells from human pluripotent stem cell (PSC)-derived neural crest cell culture by immunomagnetic positive selection.
Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated CD271+ cells, which are immediately available for downstream applications.
Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.