Microflex® Lifestar EC™ LSE-104 Nitrile Disposable Gloves
Supplier: Ansell Healthcare
Certificates
About this item
Dual-layer first responder gloves offering barrier protection against chemical hazards.
- Dual-layer, dual-color design for dual protection emergency requirements
- Specialized protection
- Non-stick/non-foaming composition for a secure, trouble-free grip
- Gloves offer advanced barrier integrity with a low 0.65 AQL
Microflex® Lifestar EC™ LSE-104 EMS gloves offer reinforced protection thanks to a dual-layer, dual-color design for first responders and healthcare workers requiring double-layered hand protection. The non-stick/non-foaming properties and textured fingertips ensure enhanced grip performance, for handling bandages, liquids, medical tape, etc. Satisfying NFPA 1999 Emergency Medical Glove Requirements and boasting chemical splash resistance certification, these nitrile exam gloves offer proven protection. The low 0.65 AQL reinforces their barrier integrity.
Certifications: ASTM D6319, EN 420:2003 + A1:2009, EN ISO 13485, FDA21 CFR 177-2600-US food contact approved, ISO 9001, NFPA 1999:2018.
Packaging: Gloves are packaged 100 per dispenser, ten dispensers per case (90 gloves per dispenser for size 3XL).
Specifications
- Catalog No:
- 76201-714
- 76201-702
- 76201-704
- 76201-706
- 76201-708
- 76201-712
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff length:
- Extended
- Extended
- Extended
- Extended
- Extended
- Extended
- Finger thickness:
- 8.7 mil
- 8.7 mil
- 8.7 mil
- 8.7 mil
- 8.7 mil
- 8.7 mil
- Palm thickness:
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- FDA regulated:
- +
- +
- +
- +
- +
- +
- EN norm:
- EN 420:2003 + A1:2009, EN ISO 13485
- EN 420:2003 + A1:2009, EN ISO 13485
- EN 420:2003 + A1:2009, EN ISO 13485
- EN 420:2003 + A1:2009, EN ISO 13485
- EN 420:2003 + A1:2009, EN ISO 13485
- EN 420:2003 + A1:2009, EN ISO 13485
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- +
- +
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +
- +
- +
- ASTM No.:
- D6319
- D6319
- D6319
- D6319
- D6319
- D6319
Specifications
Frequently Bought Together