If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
- Cat. no.:MSPP-100-0347
Specifications
About this item
Immunomagnetic isolation of untouched non-human primate B cells.
- Fast, easy to use, and column-free
- Up to 95% purity
- Untouched, viable cells
The EasySep™ Non-Human Primate B Cell Isolation Kit is designed to isolate untouched and highly purified B cells from fresh or previously frozen non-human primate peripheral blood mononuclear cells (PBMCs) in as little as 20 minutes by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction.
This kit has been verified for use with rhesus and cynomolgus macaques.