EasySep™ Human APC Positive Selection Kit II
Catalog # MSPP-17661
Supplier: STEMCELL Technologies
Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ human APC positive selection kit II
- Cat. no.:MSPP-17661
Specifications
About this item
Immunomagnetic positive selection of human cells labeled with APC-conjugated antibodies.
- Fast and easy-to-use
- No columns required
The EasySep™ Human APC Positive Selection Kit II is designed to isolate cells that are labeled with APC (allophycocyanin)-conjugated antibodies by positive selection. Desired cells are targeted with antibody complexes recognizing APC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Human APC Positive Selection Kit (STMC18451).
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).