ContactUsLinkComponent
EasySep™ FITC Positive Selection Kit II
EasySep™ FITC Positive Selection Kit II
Catalog # MSPP-17682
undefined
EasySep™ FITC Positive Selection Kit II
Catalog # MSPP-17682
Supplier Number:  17682
Specifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ FITC positive selection kit II
  • Cat. no.:
    MSPP-17682
Specifications
About this item
Immunomagnetic positive selection of any cells labeled with FITC-conjugated antibodies.
  • Fast and easy-to-use
  • No columns required
The EasySep™ FITC Positive Selection Kit II is designed to isolate cells that are labeled with FITC-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing FITC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ FITC Positive Selection Kit (STMC18558) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).