Specifications
- Capacity:1×10⁹ cells
- Description:EasySep™ human CD14 positive selection kit II
- Cat. no.:MSPP-17858
Specifications
About this item
Immunomagnetic positive selection of human CD14+ cells (e.g. monocytes).
- Fast and easy-to-use
- Up to 97% purity
- No columns required
The EasySep™ Human CD14 Positive Selection Kit II is designed to isolate highly purified CD14+ cells from fresh or previously frozen human peripheral blood mononuclear cells (PBMCs). Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD14 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD14 antigen is strongly expressed on monocytes and macrophages and weakly on granulocytes. It is also expressed on most tissue macrophages.
This product replaces the EasySep™ Human CD14 Positive Selection Kit (STMC18058), for even faster cell isolations and is compatible with all EasySep™ magnets.