Double Burette Clamp
470357-408
New Product
- Description:Double burette clamp
- Material:Cast metal
- Cat. no.:470357-408
Promotes precision and safety for your lab.
- Adjustable arms
- Cast metal construction
- Plastisol tips
- Easy to use
This sturdy clamp ensures burettes are held firm so students focus on experiments. The double burette clamp allows you to hold two burettes vertically, making it perfect for titration experiments. Whether you're demonstrating pH changes or exploring reaction rates, this clamp keeps everything stable and secure, minimizing spills and accidents.