About this item
VWR® Incubating microplate shakers feature variable-speed, microprocessor control, providing reliable, uniform mixing action with accurate temperature control and are ideal for cell cultures, immunoassays, hybridization, genetics research and bacterial suspensions.
- LCD display for temperature, speed, and time
- Includes platform ready to secure four microplates, deep well plates, or two microtube racks
- Available with clear lid or opaque lid for light sensitive samples
- RS232 interface provides two-way communication for data logging and unit control
- Durable base is easy to clean and corrosion resistant
- TUV listed for safety and electrical compliance
- Available with a NIST Traceable, ISO/IEC 17025 Calibration Certificate from Troemner
- Best-in-class, exclusive 5-year warranty
Shakers tray is designed to firmly secure plates into place with retaining springs and clips. Models available with a clear polycarbonate lid to easily view samples or with an opaque lid for light sensitive assays.
LCD display shows temperature, speed, and time for repeatable and accurate results. Easy to use control knobs allow for easy input of settings and access to the menu. Temperature resolution is in 0.1 °C increments. Speed resolution is in 1 rpm increments. Timer will display elapsed time, or, when programmed to user-defined limit up to 999 hours, 59 minutes, will count down to zero and shut off unit. Audible alarm sounds when time reaches zero.
On-board programming for eight separate, five-step programs. Recall feature retrieves the last five recently used settings.
For the best accuracy, the temperature calibration feature allows user to calibrate the temperature to an external temperature device for three separate, user defined set points.
Ramping feature slowly increases speed to set point for added safety. Triple eccentric drive system and maintenance free, brushless DC motor provide continuous duty operation and a long, reliable service life. Additional safety features include an integrated overload and unbalanced detection system, and illuminated hot warning symbols when temperatures reach 40 °C.
Unit includes a tray and all hardware for securing plates, power supply, and a 2.3 m (7.5') detachable, US three-wire power cord.
Specifications
- Catalog No:
- 77623-780
- 77623-776
- 77623-784
- 77623-788
- Speed accuracy:
- Above 100 rpm ±2% of set speed,
Below 100 rpm ±2 rpm - Above 100 rpm ±2% of set speed,
Below 100 rpm ±2 rpm - Above 100 rpm ±2% of set speed,
Below 100 rpm ±2 rpm - Above 100 rpm ±2% of set speed,
Below 100 rpm ±2 rpm
- Max. load:
- 4 plates or 2 microtube racks
- 4 plates or 2 microtube racks
- 4 plates or 2 microtube racks
- 4 plates or 2 microtube racks
- Platform W×D:
- 28×19.7 cm (11×7.75")
- 28×19.7 cm (11×7.75")
- 28×19.7 cm (11×7.75")
- 28×19.7 cm (11×7.75")
- Overall dimensions:
- 43.2×26.2×20.8 cm (17×10.3×8.2")
- 43.2×26.2×20.8 cm (17×10.3×8.2")
- 43.2×26.2×20.8 cm (17×10.3×8.2")
- 43.2×26.2×20.8 cm (17×10.3×8.2")
- Speed range:
- 100 - 1200 rpm
- 100 - 1200 rpm
- 100 - 1200 rpm
- 100 - 1200 rpm
- Orbit:
- 3 mm
- 3 mm
- 3 mm
- Electrical:
- 120 V, 3.75 A, 440 W
- 120 V, 3.75 A, 440 W
- 120 V, 3.75 A, 440 W
- 120 V, 3.75 A, 440 W