About this item
Compact Arium® Mini laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 L per day and are ideal for use in preparation of buffers, media and samples, both in life sciences applications and in analytical laboratory procedures.
Flexible: Different versions for different inlet water and connection options, as well as the desired water withdrawal
- Compact: Space-saving with a width of only 28 cm (+9 cm for versions with attached dispensing unit)
- Intuitive: Color touch display, with direct access to all important information and dispensing options
- Innovative: Depending on the type of system, our unique Bag tank technology saves time as no intensive tank cleaning with hazardous substances is required
- Reliable: Delivers consistently high water quality for reliable and reproducible results
A large, touch-activated screen and intuitive menu navigation ensure exceptionally easy operation. Regardless of your type of feed water available, use Arium® Mini, Arium® Mini Plus or Arium® Mini Plus Extend featuring our unique Bag tank technology or connect Arium® Mini Essential or Arium® Mini Essential Extend directly to your deionized water supply line in the lab.
Specifications
- Catalog No:
- CA77627-814
- CA77627-812
- CA77627-818
- CA77627-816
- Model:
- Arium® Mini Plus UV
- Arium® Mini Plus Extend
- Arium® Mini Essential UV Extend
- Arium® Mini Essential Extend
- Feed water:
- Tap water
- Tap water
- Water pretreated by reverse osmosis, distillation or deionization
- Water pretreated by reverse osmosis, distillation or deionization
- Flow rate:
- 1 L/min
- 1 L/min
- 1 L/min
- 1 L/min
- TOC content:
- <5 ppb
- <5 ppb
- ≤5 ppb
- ≤5 ppb
- Resistivity at +25 °C:
- 18.2 MΩ.cm
- 18.2 MΩ.cm
- 18.2 MΩ.cm
- 18.2 MΩ.cm
- Bacteria:
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- <1 CFU/1000 ml
- Particles:
- No particles >0.2 µm
- No particles >0.2 µm
- No particles >0.2 µm
- No particles >0.2 µm
- W×D×H:
- 280×530×510 mm
- 280×530×510 mm
- 280×530×510 mm
- 280×530×510 mm
- Weight:
- 13 kg
- 13 kg
- 13 kg
- 13 kg