The next generation Guardian 5000TM Hotplate Stirrers are engineered for safety and versatility across research, academic and industrial settings. They offer powerful heating and mixing, with premium hardware and software to support diverse users safely and reliably. Heating models reach up to 550 °C and feature three temperature control modes for quick heating or precise heating without overshoot. Select from five models with three plate sizes.
- Operation: 100 to 120 VAC or 220 to 240 VAC 50/60 Hz power entry, rotary switch knobs control
- Communication: RS232 for data logging and remote control
Applications: Temperature sensitive chemistry and biology, heat fixation, solution pH optimization, solution sterilization, endothermic reactions, denaturing of organic material, sample and solution preparation, dissolving buffers and reagents, preparing media, boiling.
Display: Backlit LCD display with white characters, integrated heating/stirring status indicators in green and a prominent hot top indicator in yellow.
Construction: Robust full metal housing, ceramic square top plate or ceramic-coated aluminum round top plate, high performance SmCo magnet.
Design features: Large dip angle front panel for easy monitoring from a distance, smooth enclosure surfaces for easy cleaning, audible alarm, IP 32 protection, rapid install splashing shield (optional).
- Catalog No:
- 77623-708
- 77623-736
- 77623-738
- 77623-734
- 77623-710
- 77623-732
- 77623-712
- 77623-714
- 77623-716
- Speed stability:
- ±2%
- —
- ±2%
- ±2%
- ±2%
- ±2%
- —
- ±2%
- ±2%
- Temperature stability:
- ±1% plate, below 100 °C ±1 °C and ±0.5% probe, below 100 °C ±0.5°C
- ±1% plate, below 100 °C ±1 °C and ±0.5% probe, below 100 °C ±0.5°C
- ±1% plate, below 100 °C ±1 °C and ±0.5% probe, below 100 °C ±0.5°C
- ±1% plate, below 100 °C ±1 °C and ±0.5% probe, below 100 °C ±0.5°C
- ±1% plate, below 100 °C ±1 °C and ±0.5% probe, below 100 °C ±0.5°C
- ±1% plate, below 100 °C ±1 °C and ±0.5% probe, below 100 °C ±0.5°C
- ±1% plate, below 100 °C ±1 °C and ±0.5% probe, below 100 °C ±0.5°C
- —
- ±1% plate, below 100 °C ±1 °C and ±0.5% probe, below 100 °C ±0.5°C
- Timer:
- 1 min - 99 h, 59 min
- 1 min - 99 h, 59 min
- 1 min - 99 h, 59 min
- 1 min - 99 h, 59 min
- 1 min - 99 h, 59 min
- 1 min - 99 h, 59 min
- 1 min - 99 h, 59 min
- 1 min - 99 h, 59 min
- 1 min - 99 h, 59 min
- Power consumption:
- 100 - 120 V, 50/60 Hz, ~8 A
- 100 - 120 V, 50/60 Hz, ~15 A
- 100 - 120 V, 50/60 Hz, ~15 A
- 100 - 120 V, 50/60 Hz, ~15 A
- 100 - 120 V, 50/60 Hz, ~15 A
- 100 - 120 V, 50/60 Hz, ~8 A
- 100 - 120 V, 50/60 Hz, ~15 A
- 100 - 120 V, 50/60 Hz, ~1 A
- 100 - 120 V, 50/60 Hz, ~15 A