Hand Generator
Hand generator
Catalog # 470355-414
Supplier: American Scientific
Supplier #: 4225-00
Jump to:
Product Details & Documents
Demonstrate the transformation of energy.
- Comes with a teacher’s guide
- Spare leads and gears sold separately
This handheld crank generator produces up to 6 volts of DC electricity and is a great power supply for many experiments while demonstrating the conversion of mechanical energy to electric energy. Other devices can be easily connected to the lead wires with crocodile clips on the front of the generator. It is housed in a tough transparent plastic so all the components can be easily observed.
Specifications
- Description:Hand generator