BASIKBench® Storage Drawers, Kewaunee®
KL447051
: Avantor
Product available on GSA Advantage®
- Color:—
- Description:Mounting bracket
- Size:—
- Cat. no.:KL447051
- Supplier No.:447051
Lockable storage drawers feature steel construction and recessed handles and can be stacked or mounted under worksurfaces.
- Drawers feature Contour™ front styling
- The box drawer is suspended with two-member ball bearing slides
- The file drawer is suspended with three-member ball bearing slides
A mounting bracket (sold separately) is required for each hanging drawer configuration that attaches directly to the worksurface. Drawers can be keyed alike, differently, by workstation, or by area.