3-Bromodibenzofuran ≥98.0% (by GC)
Fournisseur: TCI America
Certificats
À propos de cet article
Numéro CAS:26608-06-0
:
Formule:C₁₂H₇BrO
Numéro MDL:MFCD09966042
:247.09 g/mol
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Résultats des tests de spécification
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:121.0 to 125.0 deg-C
Résultats des tests de spécification
Frequently Bought Together