Order Entry
Canada
ContactUsLinkComponent
Methyl (Phenyl 5-Acetoxyacetamido-4,7,8-tri-O-acetyl-9-azido-3,5,9-trideoxy-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosid)onate ≥93.0% (by HPLC)
Methyl (Phenyl 5-Acetoxyacetamido-4,7,8-tri-O-acetyl-9-azido-3,5,9-trideoxy-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosid)onate ≥93.0% (by HPLC)
Catalog # TCM2696-100MG
Supplier:  TCI America
CAS Number:  
Methyl (Phenyl 5-Acetoxyacetamido-4,7,8-tri-O-acetyl-9-azido-3,5,9-trideoxy-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosid)onate ≥93.0% (by HPLC)
Catalog # TCM2696-100MG
Supplier:  TCI America
Supplier Number:  M2696-100MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 mg
  • Cat. no.:
    TCM2696-100MG
  • Supplier No.:
    M2696-100MG

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 93.0 area%
  • Elemental analysis (Nitrogen):
    8.50 to 9.40 %

Specification Test Results

About this item

 :
 :624.62 g/mol
Storage Temperature:Refrigerator
CAS Number:1195053-25-8
Formula:C₂₆H₃₂N₄O₁₂S