Samco™ Sedi-Pet® Disposable Transfer Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Polyethylene pipettes are used for blood bank applications, such as sedimentation procedures.
The long flexible stem can be bent to draw liquids from narrow or small volume tubes into the bulb. Pipette stems are readily heat-sealed for conversion to liquid-tight storage and transport vessels.
Specifications
- Catalog No:
- 14670-345
- 14670-180
- 14670-191
- 14670-169
- 14670-170
- 14670-196
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
Specifications
Related Information
14670-180_23100257
Conformity declaration
Thermo-Scientific Samco Transfer-Pipettes
Product Literature
Frequently Bought Together