ContactUsLinkComponent
LabExact® CFP541 Quantitative Ashless Hardened Cellulose Filter Papers, 21 to 23 µm
LabExact® CFP541 Quantitative Ashless Hardened Cellulose Filter Papers, 21 to 23 µm
New Product
Certificates
LabExact® CFP541 Quantitative Ashless Hardened Cellulose Filter Papers, 21 to 23 µm
New Product
Specifications
  • Catalog No:
  • 77590-736
  • 77590-746
  • 77590-744
  • 77590-742
  • 77590-740
  • 77590-738
  • Ash content:
  • <0.01%
  • <0.01%
  • <0.01%
  • <0.01%
  • <0.01%
  • <0.01%
  • Basis weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Retention:
  • 21 - 23 µm
  • 21 - 23 µm
  • 21 - 23 µm
  • 21 - 23 µm
  • 21 - 23 µm
  • 21 - 23 µm