ContactUsLinkComponent
LabExact® CFP540 Quantitative Ashless Hardened Cellulose Filter Paper, 7 - 8 µm
New Product
LabExact® CFP540 Quantitative Ashless Hardened Cellulose Filter Paper, 7 - 8 µm
New Product
  • Catalog No:
  • 77590-734
  • 77590-732
  • 77590-730
  • 77590-728
  • 77590-726
  • 77590-724
  • Ash content:
  • <0.01%
  • <0.01%
  • <0.01%
  • <0.01%
  • <0.01%
  • <0.01%
  • Basis weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Retention:
  • 7 - 8 µm
  • 7 - 8 µm
  • 7 - 8 µm
  • 7 - 8 µm
  • 7 - 8 µm
  • 7 - 8 µm
Frequently Bought Together