LabExact® CFP1 Qualitative Cellulose Filter Papers, 11 µm
Supplier: I.W. TREMONT CO., INC.
New Product
Certificates
About this item
Very widely used filter media demonstrating retention of approximately 11 µm and medium flow rate.
- Consistent bright white color of this media is ideal for photometric stain intensity measurement of air samples
- Media can also be impregnated with reagents for use in quantifying optical reflectance in gas detection procedures
Used in a broad range of laboratory and environmental applications, this media is ideally suited in separating lead sulfate, calcium carbonate, and calcium oxalate precipitates. This media is the standard for agricultural procedures such as soil and seed sample testing. It is commonly used as a separation media in the food and beverage industry to extract liquids from solid samples.
Packaging: Box of 100 filters.
Specifications
- Catalog No:
- 77590-530
- 77590-528
- 77590-526
- 77590-524
- 77590-522
- 77590-520
- 77590-518
- 77590-516
- 77590-532
- 77590-514
- Ash content:
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- 0.05%
- Basis weight:
- 86 g/m²
- 86 g/m²
- 86 g/m²
- 86 g/m²
- 86 g/m²
- 86 g/m²
- 86 g/m²
- 86 g/m²
- 86 g/m²
- 86 g/m²
- Retention:
- 11 µm
- 11 µm
- 11 µm
- 11 µm
- 11 µm
- 11 µm
- 11 µm
- 11 µm
- 11 µm
- 11 µm
Specifications
Frequently Bought Together