Orders LinkContactUsLinkComponent
LabExact® Extraction Thimbles, Cellulose
LabExact® Extraction Thimbles, Cellulose
LabExact® Extraction Thimbles, Cellulose
Certificates
LabExact® Extraction Thimbles, Cellulose
LabExact® Extraction Thimbles, Cellulose
53 Options Available Below
Product Details & Documents
LabExact® high quality cellulose extraction thimbles are available in a variety of dimensions to fit most Soxhlet extraction units.
  • Extraction thimbles are manufactured with high purity cellulose fibers and have a consistent wall thickness
  • Available as fast flow, with lid and extra density
Common applications include air and waste gas analysis, collection of solid particles such as dust, concrete slurry evaluation, analysis of pesticide residues, oil/fat content of solid foods and oil and grease analysis of solid wastes.
Documents
Product Family Options
Product Information
  • Material
    I.D.×H
    Wall thickness
    Particle retention
    Availability
    Price
  • 77590-784
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    8×40 mm
    1.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5001
  • 77590-786
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    9×50 mm
    1.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5002
  • 77590-788
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    15×50 mm
    1.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5003
  • 77590-790
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    15×100 mm
    1.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5004
  • 77590-792
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    20×80 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $195.05
    $195.05
    Box of 25
       $195.05
    Supplier Number: LECT5005
  • 77590-794
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    22×80 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $195.05
    $195.05
    Box of 25
       $195.05
    Supplier Number: LECT5006
  • 77590-796
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    23×90 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5007
  • 77590-798
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    23×100 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5008
  • 77590-800
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    27×80 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5009
  • 77590-802
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    27×100 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5010
  • 77590-804
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    27×60 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5011
  • 77590-806
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    28×100 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5013
  • 77590-808
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    28×120 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $209.93
    $209.93
    Box of 25
       $209.93
    Supplier Number: LECT5014
  • 77590-810
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    28×80 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5015
  • 77590-812
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    28×90 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5016
  • 77590-814
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    29×100 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $209.93
    $209.93
    Box of 25
       $209.93
    Supplier Number: LECT5017
  • 77590-816
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    30×150 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $265.48
    $265.48
    Box of 25
       $265.48
    Supplier Number: LECT5018
  • 77590-818
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    30×60 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5019
  • 77590-820
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    30×80 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $165.45
    $165.45
    Box of 25
       $165.45
    Supplier Number: LECT5020
  • 77590-822
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    30×90 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $218.63
    $218.63
    Box of 25
       $218.63
    Supplier Number: LECT5021
  • 77590-824
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    33×94 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $235.90
    $235.90
    Box of 25
       $235.90
    Supplier Number: LECT5022
  • 77590-826
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    30×100 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $221.98
    $221.98
    Box of 25
       $221.98
    Supplier Number: LECT5023
  • 77590-828
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    31×118 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $262.33
    $262.33
    Box of 25
       $262.33
    Supplier Number: LECT5024
  • 77590-830
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    31×130 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $283.05
    $283.05
    Box of 25
       $283.05
    Supplier Number: LECT5025
  • 77590-832
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    33×205 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $355.73
    $355.73
    Box of 25
       $355.73
    Supplier Number: LECT5026
  • 77590-834
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    34×120 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $262.33
    $262.33
    Box of 25
       $262.33
    Supplier Number: LECT5027
  • 77590-836
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    34×150 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $308.86
    $308.86
    Box of 25
       $308.86
    Supplier Number: LECT5028
  • 77590-838
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    38×200 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $413.86
    $413.86
    Box of 25
       $413.86
    Supplier Number: LECT5029
  • 77590-840
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    40×150 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $340.33
    $340.33
    Box of 25
       $340.33
    Supplier Number: LECT5030
  • 77590-842
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    40×123 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $306.82
    $306.82
    Box of 25
       $306.82
    Supplier Number: LECT5031
  • 77590-844
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    43×130 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $335.22
    $335.22
    Box of 25
       $335.22
    Supplier Number: LECT5032
  • 77590-846
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    48×145 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $400.86
    $400.86
    Box of 25
       $400.86
    Supplier Number: LECT5033
  • 77590-848
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    48×200 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $469.42
    $469.42
    Box of 25
       $469.42
    Supplier Number: LECT5034
  • 77590-850
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    48×230 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $472.30
    $472.30
    Box of 25
       $472.30
    Supplier Number: LECT5035
  • 77590-852
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    51×145 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $451.87
    $451.87
    Box of 25
       $451.87
    Supplier Number: LECT5036
  • 77590-854
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    51×180 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $481.05
    $481.05
    Box of 25
       $481.05
    Supplier Number: LECT5037
  • 77590-856
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    57×315 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $714.26
    $714.26
    Box of 25
       $714.26
    Supplier Number: LECT5038
  • 77590-858
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    60×180 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $472.30
    $472.30
    Box of 25
       $472.30
    Supplier Number: LECT5039
  • 77590-860
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    55×275 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $597.63
    $597.63
    Box of 25
       $597.63
    Supplier Number: LECT5040
  • 77590-862
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    75×160 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $539.33
    $539.33
    Box of 25
       $539.33
    Supplier Number: LECT5041
  • 77590-864
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    68×250 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $641.36
    $641.36
    Box of 25
       $641.36
    Supplier Number: LECT5042
  • 77590-866
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    70×330 mm
    2.0 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $787.14
    $787.14
    Box of 25
       $787.14
    Supplier Number: LECT5043
  • 77590-888
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    33×80 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $182.82
    $182.82
    Box of 25
       $182.82
    Supplier Number: LECT5951
  • 77590-868
    LabExact® Extraction thimbles cellulose, standard format
    LabExact® Extraction thimbles cellulose, standard format
    Cotton linter cellulose
    90×200 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $932.75
    $932.75
    Box of 25
       $932.75
    Supplier Number: LECT5055
  • 77590-886
    LabExact® Extraction thimbles cellulose, collar and recess format
    LabExact® Extraction thimbles cellulose, collar and recess format
    Cotton linter cellulose
    79×155 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $787.14
    $787.14
    Box of 25
       $787.14
    Supplier Number: LECT5500
  • 77590-882
    LabExact® Extraction thimbles cellulose, extra density format
    LabExact® Extraction thimbles cellulose, extra density format
    Cotton linter cellulose
    22×80 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5406
  • 77590-884
    LabExact® Extraction thimbles cellulose, extra density format
    LabExact® Extraction thimbles cellulose, extra density format
    Cotton linter cellulose
    30×100 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $218.63
    $218.63
    Box of 25
       $218.63
    Supplier Number: LECT5423
  • 77590-870
    LabExact® Extraction thimbles cellulose, fast flow format
    LabExact® Extraction thimbles cellulose, fast flow format
    Cotton linter cellulose
    20×80 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5105
  • 77590-872
    LabExact® Extraction thimbles cellulose, fast flow format
    LabExact® Extraction thimbles cellulose, fast flow format
    Cotton linter cellulose
    28×90 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $180.77
    $180.77
    Box of 25
       $180.77
    Supplier Number: LECT5116
  • 77590-874
    LabExact® Extraction thimbles cellulose, fast flow format
    LabExact® Extraction thimbles cellulose, fast flow format
    Cotton linter cellulose
    31×205 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $355.73
    $355.73
    Box of 25
       $355.73
    Supplier Number: LECT5126
  • 77590-876
    LabExact® Extraction thimbles cellulose, with lid
    LabExact® Extraction thimbles cellulose, with lid
    Cotton linter cellulose
    30×80 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $310.97
    $310.97
    Box of 25
       $310.97
    Supplier Number: LECT5220
  • 77590-878
    LabExact® Extraction thimbles cellulose, with lid
    LabExact® Extraction thimbles cellulose, with lid
    Cotton linter cellulose
    30×100 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $364.40
    $364.40
    Box of 25
       $364.40
    Supplier Number: LECT5223
  • 77590-880
    LabExact® Extraction thimbles cellulose, with lid
    LabExact® Extraction thimbles cellulose, with lid
    Cotton linter cellulose
    31×130 mm
    1.5 mm
    5 µm
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $428.62
    $428.62
    Box of 25
       $428.62
    Supplier Number: LECT5225