Stainless Steel Worktables, Advance Tabco®
Supplier: Advance Tabco
Certificates
About this item
These 16-gauge, Type 304 stainless steel tables provide a sturdy work area in the cleanroom.
- Rounded front and rear edges, and the sides are turned down on a 90° angle for easy alignment with other equipment
- Reinforced with stainless steel hat channels to maintain a level working surface
- 4.1 cm (1⁵/₈") diameter table legs
- 76 cm (30") working height with height adjustment 2.5 cm (1")
All-stainless steel construction resists corrosion and helps eliminate contamination concerns. The fasteners used for the installation of the horizontal leg bar are also constructed of stainless steel.
Ordering information: Units can be electropolished for an additional charge; contact your Avantor Furniture Specialist for more information.
Customers Who Bought This Also Bought