Order Entry
Canada
ContactUsLinkComponent
LabExact® Grade F 0.7 µm Binderless Glass Microfiber Filters
LabExact® Grade F 0.7 µm Binderless Glass Microfiber Filters
New Product
Certificates
undefined
LabExact® Grade F 0.7 µm Binderless Glass Microfiber Filters
New Product

Specifications

  • Catalog No:
  • 77591-180
  • 77591-178
  • 77591-176
  • 77591-174
  • 77591-172
  • 77591-170
  • 77591-168
  • 77591-166
  • 77591-164
  • 77591-162
  • 77591-160
  • 77591-158
  • 77591-156
  • 77591-182
  • Water flow rate:
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • 80 s/100 ml, 47 mm Ø @ 10 mbar
  • Airflow rate:
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • 19 s/100 ml/sq. in.
  • Retention:
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm
  • 0.7 µm

Specifications

Frequently Bought Together