Ceiling-Mount Emergency Showers, WaterSaver Faucet
Supplier: WaterSaver Faucet
Certificates
About this item
Emergency showers provide a spray pattern 50.8 cm (20") in Ø at 152.4 cm (60") above the floor.
Featuring a stainless steel pull rod and stay-open ball valve, the showers are available in two styles: pendant and flush. The pendant-style shower has a 25.4 cm (10") stainless steel shower head, nipple, escutcheon, and ceiling guide plate. The flush-style shower has a 25.4 cm (10") white epoxy/polyester hybrid powder-coated cast brass shower head. Both showers are furnished with a 2.5 cm (1") IPS female inlet.
Certifications: Meet ANSI Z358.1 standard.
Frequently Bought Together