Eyewash System Combination Eyewash/Sink Faucet, Speakman®
Catalog # 83008-084
Supplier: Speakman
Small Business Enterprise
Specifications
- Description:Eyewash System* Combination Eyewash/Sink Faucet
- Cat. no.:83008-084
- Supplier No.:SEF-9000
Specifications
About this item
System combines an eyewash/drench hose with a service sink faucet.
This creative design allows the eyewash to be attached directly to the faucet’s cold water integral stop cavity and permits both units to be operated independently. As a result, only one plumbing connection is required and there is never a worry about delivering scalding water to the eyes.
Certifications: Meets ANSI Z358.1 standard.