Wall-Mount Swing-Down Eyewash Station, Barrier-Free, Speakman®
Catalog #: 89027-704
Supplier: Speakman
Small Business Enterprise
Specifications
- Description:Wall-Mount Swing-Down Eyewash Station
- Cat. no.:89027-704
- Supplier No.:SE-575SD-ADA
Specifications
About this item
These eyewash stations are designed to provide convenient and immediate flushing capabilities to workers in a variety of environments.
- Feature chrome-plated head, arms, and valves
- High-visibility signs are included for identification and activation instructions
Stations are encased in a stainless steel cabinet that is installed flush with the wall to keep corridors free from obstruction. Units are activated automatically by pulling spray arms downward 90°. Water will continue to discharge from the spray outlets until the arms are manually pushed upright into the off position. A lockable access panel allows for easy serviceability from the front of the unit. This eyewash station is ADA-compliant, and features extended arms that provide more convenient accessibility for persons with physical disabilities.
Certifications: Meets ANSI Z358.1 standard.
Customers Who Bought This Also Bought