Order Entry
Canada
Orders LinkContactUsLinkComponent
Portable Table, Labconco®

Portable Table, Labconco®

Portable table, Steel, 36⅜", 34⅞×19", 44 lbs.
Catalog # 82006-866
Supplier:   Labconco
Supplier #:  8025000
Portable Table, Labconco®
Catalog # 82006-866
Supplier:  Labconco
Portable Table, Labconco®
Portable Table, Labconco®

Portable table, Steel, 36⅜", 34⅞×19", 44 lbs.

Price
$1,200.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Safely transports heavy instruments and bulky equipment.

  • Shelves are chemical- and heat-resistant phenolic board with high-pressure melamine laminate surface

Holds up to 181 kg (400 lbs.). Welded frame is constructed of rugged, 18-gauge glacier white epoxy-coated 2.5 cm (1") square tubular steel. Cart features four 10.2 cm (4") diameter casters (two with toe locks) and two handles with ergonomic extruded rubber grips.

Ordering information: Manufacturer's lifetime warranty. Shipped fully assembled.

Documents

Specifications
  • Worksurface material:
    Steel
  • Description:
    Portable table
  • Height:
    36⅜"
  • W×D:
    34⅞×19"
  • Shipping weight:
    44 lbs.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.