PYREX® Disposable Milk Pipettes, Borosilicate Glass, Sterile, Corning
Catalog # 89091-228
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Capacity:2.2 ml
- Environmentally Preferable:
- Cat. no.:89091-228
- Subdivisions:1.0, 2.0, 2.1, 2.2 ml
- Supplier No.:7058-2X
Specifications
About this item
These borosilicate glass pipettes meet requirements recommended by APHA as shown in 'Standard Methods for the Examination of Dairy Products.' Pipette is convenient when duplicate platings are to be made on the same medium for determination of gram-negative bacteria, mold and yeast, or total plate count.
- Blow-out is not required
- ISO color-coding is printed on the tear strip for easy identification
Note: Raw milk products may contain pathogenic microorganisms. For user safety, a mechanical pipetting device is recommended.
Ordering information: Packaged in sterile bags, 25 pieces per bag.