Order Entry
Canada
Orders LinkContactUsLinkComponent
StatSpin® Express 2 Centrifuge, Iris Sample Processing

StatSpin® Express 2 Centrifuge, Iris Sample Processing

Centrifuge
Catalog # CA56430-040
Supplier:   Statspin
Supplier #:  SSX2
StatSpin® Express 2 Centrifuge, Iris Sample Processing
Catalog # CA56430-040
Supplier:  Statspin
StatSpin® Express 2 Centrifuge, Iris Sample Processing
StatSpin® Express 2 Centrifuge, Iris Sample Processing

Centrifuge

Price
$7,276.55
Log in for availability
Each
Product Details & Documents

With high productivity, improved safety, and lower cost per test, this centrifuge is the perfect solution to sample prep bottlenecks in a busy clinical lab.

  • Two-Minute Automatic Cycle
  • Rapid Deceleration
  • Quiet
  • Compact
  • No Waiting for Batches

It's compact and quiet, making it ideal for use right at the workstation

The two-minute automatic cycle, combined with rapid, gentle deceleration, allows the processing of tubes as they arrive, with no waiting for batches. The centrifuge accepts standard glass or plastic blood collection tubes up to 5 mL (13x75 mm). The unique rotor design minimizes the chance of breakage.

It spins standard 5 mL blood collection tubes in only two minutes, yet it produces quality serum or plasma equal to 10–15 minute runs in larger, more costly equipment.

Certifications: UL listed. CE marked.

Ordering information: UL listed. CE marked.

Caution: Note: StatSpin centrifuges have a 6 month warranty.

Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.

Specifications
  • Description:
    Centrifuge
  • Cat. No.:
    CA56430-040
  • Max. speed:
    8500 rpm
  • Acceleration/braking time:
    20 s (to 95% of speed) / <30 second s
  • Dimensions:
    15.2H x 16.8W x 21.8D cm (6 x 619/32 x 819/32")
  • Electrical:
    120–240V, 50/60Hz
  • Max. RCF:
    4440 g
  • Cycle time:
    30, 120, 180 s
  • Weight:
    2.3 kg (5 lbs.)
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Inserts for 4–5 mL (13 x 75 mm, 10.25 x 68 mm) Tubes
    Inserts for 4–5 mL (13 x 75 mm, 10.25 x 68 mm) Tubes
    CA56430-142
    Log in for availability
    Pack of 4 - $82.11

Product Information

  • Catalog #
    Availability
    Price
  • Disposable Bowl Liners
    Disposable Bowl Liners
    CA56430-140
    Log in for availability
    Pack of 10 - $703.86

Product Information

  • Catalog #
    Availability
    Price
  • Inserts for 3 mL (10.25 x 64 mm) Tubes
    Inserts for 3 mL (10.25 x 64 mm) Tubes
    CA56430-144
    Log in for availability
    Pack of 4 - $80.65