Order Entry
Canada
ContactUsLinkComponent
 
LabExact® Grade A-E 1.0 µm Binderless Glass Microfiber Filters
New Product
 
undefined
LabExact® Grade A-E 1.0 µm Binderless Glass Microfiber Filters
New Product

 

  • Catalog No:
  • 77590-398
  • 77590-396
  • 77590-406
  • 77590-414
  • 77590-412
  • 77590-410
  • 77590-408
  • 77590-394
  • 77590-392
  • 77590-390
  • 77590-388
  • 77590-404
  • 77590-402
  • 77590-400
  • Water flow rate:
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • 15 s/100 ml, 47 mm Ø @ 10 mbar
  • Airflow rate:
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • 6 s/100 ml/sq. in.
  • Retention:
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm
  • 1.0 µm

 

Frequently Bought Together