Granite Platform with Anti-Vibration Isolation
Supplier: Sartorius
Certificates
About this item
These Anti-vibration granite platforms with vibration isolators have specially designed 1.5" thick silicone vibration isolators that are connected to the platform at the base and help to eliminate vibration and enhance balance response time.
Extended model is ideal for larger balances. Key features Ideal for use with any analytical, semi-micro, micro, and ultra-micro balances.
Frequently Bought Together