Essential Weights™ Calibration Weight Sets, ASTM Class 0 and 1
Supplier: Sartorius
Certificates
About this item
Essential Weights are customized sets of three or four stainless steel test weights in a small, durable carrying case, specifically tailored to your precision weighing equipment.
- ASTM Class 0 and Class 1 weights
- Polished stainless steel
- Density is approximately 7.95 g/cm³
- Special PVC weight case
- Configurations available ideal for calibration, shift tests and sensitivity tests
- Other configurations available by special order
- Meets USP-41 requirements for intermediate checks
Packaging: Includes: Precision tweezers, NVLAP Accredited Certificate of Calibration.
Frequently Bought Together